Gene Transcripts

Gene Symbol
Gtpbp4
Entrez Gene ID
69237
Gene Name
GTP binding protein 4
Gene Aliases
2610028C09Rik, Crfg, Gtpbp3, NGB, Nog1
Chromosome Location
Chr. 13: 8972464 - 8996012 on Build GRCm38
UniGene ID
Mm.41800
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027000.4 NP_081276.2 4 464
GenBank mRNA AF325354.1 - 4 448
AF348208.1 - 4 471
AK011580.1 - 4 475
AK141852.1 - 4 474
AK163372.1 - 3 271
AK164330.1 - 4 465
AK166711.1 - 4 470
BC005514.1 - 4 445
BC025431.1 - 4 469 5007174

View Full Product Details