Gene Transcripts

Gene Symbol
Chmp4c
Entrez Gene ID
66371
Gene Name
charged multivesicular body protein 4C
Gene Aliases
2010012P02Rik, 2210015K02Rik, 2310010I16Rik, Shax3, Snf7-3
Chromosome Location
Chr. 3: 10366973 - 10391005 on Build GRCm38
UniGene ID
Mm.366147
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025519.2 NP_079795.1 5 1336
GenBank mRNA AK009273.1 - 5 1338
AK033502.1 - 229
BC006951.1 - 191
BC115983.1 - 5 1322 40105850
BC117538.1 - 5 1326 40105849

View Full Product Details