Gene Transcripts
- Gene Symbol
- Morf4l1
- Entrez Gene ID
- 21761
- Gene Name
- mortality factor 4 like 1
- Gene Aliases
- MORFRG15, MRG15, TEG-189, Tex189, mKIAA4002
- Chromosome Location
- Chr. 9: 90091669 - 90114820 on Build GRCm38
- UniGene ID
- Mm.426209
- Species
- Mus musculus
- Gene Symbol
- Gm7420
- Entrez Gene ID
- 664949
- Gene Name
- predicted gene 7420
- Gene Aliases
- EG664949
- Chromosome Location
- Chr. 5: 29217629 - 29218650 on Build GRCm38
- UniGene ID
- Mm.442567
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK141442.1 |
- |
|
3634 |
|
View Full Product Details