Gene Transcripts

Gene Symbol
Zfp52
Entrez Gene ID
22710
Gene Name
zinc finger protein 52
Gene Aliases
KRAB11, Zfp-52, Zfp76, mszf23-2, mszf24, mszf34, mszf75, zfec29
Chromosome Location
Chr. 17: 21516295 - 21562601 on Build GRCm38
UniGene ID
Mm.296100
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144515.2 NP_653098.2 5 2550
GenBank mRNA AB010337.1 - 1 124
AK032299.1 - 5 2551
BC021527.1 - 5 2462 5353641
M74452.1 - 1 41

View Full Product Details