Gene Transcripts
- Gene Symbol
- Mpp4
- Entrez Gene ID
- 58808
- Gene Name
- membrane protein, palmitoylated 4 (MAGUK p55 subfamily member 4)
- Gene Aliases
- Dlg6
- Chromosome Location
- Chr. 9: 65917917 - 65961267 on Build Rnor_6.0
- UniGene ID
- Rn.163075
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB030499.1 |
- |
4, 5 |
527 |
|
AB030500.1 |
- |
4, 5 |
527 |
|
AB030501.1 |
- |
4, 5 |
527 |
|
View Full Product Details