Gene Transcripts

Gene Symbol
Guca2b
Entrez Gene ID
64055
Gene Name
guanylate cyclase activator 2B
Gene Aliases
Chromosome Location
Chr. 5: 138695591 - 138697641 on Build Rnor_6.0
UniGene ID
Rn.97593
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022284.2 NP_071620.1 2 140
GenBank mRNA U41322.1 - 2 140
U73898.1 - 2 156
U75186.1 - 2 267

View Full Product Details