Gene Transcripts

Gene Symbol
Samsn1
Entrez Gene ID
170637
Gene Name
SAM domain, SH3 domain and nuclear localization signals, 1
Gene Aliases
Nash
Chromosome Location
Chr. 11: 14254047 - 14304603 on Build Rnor_6.0
UniGene ID
Rn.81187
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130821.1 NP_570834.1 3 176
GenBank mRNA AB067679.1 - 3 176

View Full Product Details