Gene Transcripts

Gene Symbol
Rhoq
Entrez Gene ID
85428
Gene Name
ras homolog family member Q
Gene Aliases
Tc10
Chromosome Location
Chr. 6: 10533151 - 10568582 on Build Rnor_6.0
UniGene ID
Rn.4169
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053522.1 NP_445974.1 3 617
GenBank mRNA AB031482.1 - 3 617
BC061760.1 - 3 607 5621223

View Full Product Details