Gene Transcripts

Gene Symbol
Plaa
Entrez Gene ID
116645
Gene Name
phospholipase A2, activating protein
Gene Aliases
PLA2P, Plap
Chromosome Location
Chr. 5: 113548913 - 113578928 on Build Rnor_6.0
UniGene ID
Rn.7441
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053866.3 NP_446318.3 2 394
GenBank mRNA CK366206.1 - 2 394
FQ211509.1 - 455
FQ223782.1 - 2 469
U17901.1 - 2 253

View Full Product Details