Gene Transcripts

Gene Symbol
Serping1
Entrez Gene ID
295703
Gene Name
serpin peptidase inhibitor, clade G (C1 inhibitor), member 1
Gene Aliases
Chromosome Location
Chr. 3: 72161230 - 72171109 on Build Rnor_6.0
UniGene ID
Rn.100285
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199093.1 NP_954524.1 2 274
XM_006234447.2 XP_006234509.1 3 349
GenBank mRNA BC061860.1 - 2 274

View Full Product Details