Gene Transcripts

Gene Symbol
Cyp20a1
Entrez Gene ID
316435
Gene Name
cytochrome P450, family 20, subfamily a, polypeptide 1
Gene Aliases
Chromosome Location
Chr. 9: 67130259 - 67179693 on Build Rnor_6.0
UniGene ID
Rn.101629
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199401.1 NP_955433.1 3 305
GenBank mRNA BC061716.1 - 3 305

View Full Product Details