Gene Transcripts

Gene Symbol
Cntn2
Entrez Gene ID
25356
Gene Name
contactin 2 (axonal)
Gene Aliases
TAG-1, TAG-564, TAG1, Tax
Chromosome Location
Chr. 13: 49280913 - 49314047 on Build Rnor_6.0
UniGene ID
Rn.89910
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012884.1 NP_037016.1 3 334
XM_006249746.2 XP_006249808.1 4 548
XM_008769427.1 XP_008767649.1 3 437
GenBank mRNA M31725.1 - 3 334

View Full Product Details