Gene Transcripts

Gene Symbol
Gata6
Entrez Gene ID
29300
Gene Name
GATA binding protein 6
Gene Aliases
Chromosome Location
Chr. 18: 2416871 - 2446338 on Build Rnor_6.0
UniGene ID
Rn.8701
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019185.1 NP_062058.1 2 907
GenBank mRNA L22760.1 - 2 907

View Full Product Details