Gene Transcripts

Gene Symbol
Adam7
Entrez Gene ID
29641
Gene Name
ADAM metallopeptidase domain 7
Gene Aliases
7, ADAM, ADAM 7, EAP, EAP I, EAPI, I
Chromosome Location
Chr. 15: 49418946 - 49467174 on Build Rnor_6.0
UniGene ID
Rn.10357
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020301.1 NP_064697.1 3, 4 283
GenBank mRNA X66140.1 - 3, 4 283

View Full Product Details