Gene Transcripts

Gene Symbol
Slc9a3
Entrez Gene ID
24784
Gene Name
solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3
Gene Aliases
Nhe3
Chromosome Location
Chr. 1: 31734327 - 31777158 on Build Rnor_6.0
UniGene ID
Rn.9706
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012654.1 NP_036786.1 3 690
XM_008758677.1 XP_008756899.1 3 778
GenBank mRNA M85300.1 - 3 690

View Full Product Details