Gene Transcripts
- Gene Symbol
- Adra2b
- Entrez Gene ID
- 24174
- Gene Name
- adrenoceptor alpha 2B
- Gene Aliases
- Chromosome Location
- Chr. 3: 119805941 - 119809987 on Build Rnor_6.0
- UniGene ID
- Rn.10296
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138505.2 |
NP_612514.2
|
1 |
1874 |
|
GenBank mRNA |
M32061.1 |
- |
1 |
1871 |
|
View Full Product Details