Gene Transcripts

Gene Symbol
SERTAD1
Entrez Gene ID
29950
Gene Name
SERTA domain containing 1
Gene Aliases
SEI1, TRIP-Br1
Chromosome Location
Chr. 19: 40422501 - 40426025 on Build GRCh38
UniGene ID
Hs.269898
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013376.3 NP_037508.2 2 680
GenBank mRNA AF117959.1 - 2 651
AF366402.1 - 2 632
AK074652.1 - 2 680
AK222970.1 - 2 647
AK223011.1 - 2 647
BC002670.2 - 2 629 3613213

View Full Product Details