Gene Transcripts

Gene Symbol
COPS5
Entrez Gene ID
10987
Gene Name
COP9 signalosome subunit 5
Gene Aliases
CSN5, JAB1, MOV-34, SGN5
Chromosome Location
Chr. 8: 67043079 - 67062327 on Build GRCh38
UniGene ID
Hs.491912
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006837.2 NP_006828.2 5 934
GenBank mRNA AB209134.1 - 4 2644
AK056967.1 - 7 710
AK128519.1 - 4 2926
AK310434.1 - 5 736
AK311071.1 - 5 751
BC001187.1 - 5 738 3354130
BC001859.2 - 5 716 3530890
BC007272.1 - 5 712 3161025
BX648542.1 - 5 3271
CR541678.1 - 5 603
U65928.1 - 5 730
U70734.1 - 5 723

View Full Product Details