Gene Transcripts

Gene Symbol
TOMM7
Entrez Gene ID
54543
Gene Name
translocase of outer mitochondrial membrane 7
Gene Aliases
TOM7
Chromosome Location
Chr. 7: 22812632 - 22822852 on Build GRCh38
UniGene ID
Hs.112318
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019059.3 NP_061932.1 3 705
GenBank mRNA BC071973.1 - 3 655

View Full Product Details