Gene Transcripts

Gene Symbol
Them6
Entrez Gene ID
300015
Gene Name
thioesterase superfamily member 6
Gene Aliases
RGD1359378
Chromosome Location
Chr. 7: 115956276 - 115959453 on Build Rnor_6.0
UniGene ID
Rn.98281
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007658.1 NP_001007659.1 2 644
GenBank mRNA BC083742.1 - 2 644

View Full Product Details