Gene Transcripts

Gene Symbol
Olr292
Entrez Gene ID
293594
Gene Name
olfactory receptor 292
Gene Aliases
ratchr1-200419540-200420457_ORF
Chromosome Location
Chr. 1: 212885464 - 212886381 on Build Rnor_6.0
UniGene ID
Rn.141742
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000232.1 NP_001000232.1 1 705

View Full Product Details