Gene Transcripts

Gene Symbol
Mrgprd
Entrez Gene ID
293648
Gene Name
MAS-related GPR, member D
Gene Aliases
Mgrd, Mrgd, TGR7
Chromosome Location
Chr. 1: 218481148 - 218489515 on Build Rnor_6.0
UniGene ID
Rn.162482
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001506.1 NP_001001506.1 1 115
XM_006230725.2 XP_006230787.1 2 1034
GenBank mRNA AB154411.1 - 1 115

View Full Product Details