Gene Transcripts
- Gene Symbol
- PLCB1
- Entrez Gene ID
- 23236
- Gene Name
- phospholipase C beta 1
- Gene Aliases
- EIEE12, PI-PLC, PLC-154, PLC-I, PLC-beta-1, PLC154, PLCB1A, PLCB1B
- Chromosome Location
- Chr. 20: 8132236 - 8884911 on Build GRCh38
- UniGene ID
- Hs.431173
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015192.3 |
NP_056007.1
|
2, 3 |
572 |
|
NM_182734.2 |
NP_877398.1
|
2, 3 |
572 |
|
XM_011529199.2 |
XP_011527501.1
|
2, 3 |
532 |
|
XM_017027752.1 |
XP_016883241.1
|
2, 3 |
532 |
|
XM_017027753.1 |
XP_016883242.1
|
2, 3 |
159 |
|
GenBank mRNA |
AB011153.3 |
- |
2, 3 |
572 |
|
AJ278313.1 |
- |
2, 3 |
188 |
|
AJ278314.1 |
- |
2, 3 |
188 |
|
AK299195.1 |
- |
2, 3 |
207 |
|
AK310597.1 |
- |
3, 4 |
500 |
|
AY004175.1 |
- |
2, 3 |
170 |
|
BC069420.1 |
- |
2, 3 |
188 |
7262405 |
BC117231.1 |
- |
2, 3 |
243 |
40125782 |
View Full Product Details