Gene Transcripts
- Gene Symbol
- AAAS
- Entrez Gene ID
- 8086
- Gene Name
- aladin WD repeat nucleoporin
- Gene Aliases
- AAA, AAASb, ADRACALA, ADRACALIN, ALADIN, GL003
- Chromosome Location
- Chr. 12: 53307456 - 53321628 on Build GRCh38
- UniGene ID
- Hs.369144
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001173466.1 |
NP_001166937.1
|
3, 4 |
477 |
|
NM_015665.5 |
NP_056480.1
|
3, 4 |
477 |
|
XM_011538778.1 |
XP_011537080.1
|
3, 4 |
409 |
|
XM_011538780.1 |
XP_011537082.1
|
3, 4 |
409 |
|
GenBank mRNA |
AF226048.1 |
- |
3, 4 |
396 |
|
AJ289857.1 |
- |
3, 4 |
404 |
|
AK000833.1 |
- |
3, 4 |
404 |
|
AK094302.1 |
- |
2, 3 |
756 |
|
AK222509.1 |
- |
3, 4 |
404 |
|
AK293342.1 |
- |
3, 4 |
434 |
|
AY237818.1 |
- |
3, 4 |
445 |
|
BC000659.2 |
- |
3, 4 |
439 |
3347423 |
BC018836.2 |
- |
3, 4 |
439 |
|
BT006912.1 |
- |
3, 4 |
314 |
|
DC393722.1 |
- |
3, 4 |
477 |
|
View Full Product Details