Gene Transcripts

Gene Symbol
GDF2
Entrez Gene ID
2658
Gene Name
growth differentiation factor 2
Gene Aliases
BMP-9, BMP9, HHT5
Chromosome Location
Chr. 10: 47322490 - 47326270 on Build GRCh38
UniGene ID
Hs.279463
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016204.2 NP_057288.1 1 471
GenBank mRNA AF188285.1 - 1 455
AK314956.1 - 1 471
BC069643.1 - 1 374 7262433
BC074921.2 - 1 323 30915569

View Full Product Details