Gene Transcripts

Gene Symbol
TBL1Y
Entrez Gene ID
90665
Gene Name
transducin (beta)-like 1, Y-linked
Gene Aliases
TBL1
Chromosome Location
Chr. Y: 6910686 - 7107154 on Build GRCh38
UniGene ID
Hs.664560
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033284.1 NP_150600.1 8 1052
NM_134258.1 NP_599020.1 7 1021
NM_134259.1 NP_599021.1 7 957
XM_005262572.3 XP_005262629.1 2 716
XM_017030086.1 XP_016885575.1 8 1041
XM_017030087.1 XP_016885576.1 8 1041
GenBank mRNA AF332220.1 - 8 1052
AF332221.1 - 7 1021
AF332222.1 - 7 957
AK298210.1 - 7 933
BC130471.1 - 4 535 40146502
BC130473.1 - 4 535 40146504

View Full Product Details