Gene Transcripts
- Gene Symbol
- Mtus1
- Entrez Gene ID
- 102103
- Gene Name
- mitochondrial tumor suppressor 1
- Gene Aliases
- AI481402, ATBP135, Atip1, B430010I23Rik, B430305I03Rik, C85752, Cctsg1-440, MD44, MTSG1, mKIAA1288
- Chromosome Location
- Chr. 8: 40990912 - 41133983 on Build GRCm38
- UniGene ID
- Mm.149438
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001005863.2 |
NP_001005863.1
|
8, 9 |
3342 |
|
NM_001005864.3 |
NP_001005864.1
|
6, 7 |
948 |
|
NM_001005865.3 |
NP_001005865.2
|
4, 5 |
961 |
|
NM_001286413.1 |
NP_001273342.1
|
8, 9 |
3224 |
|
GenBank mRNA |
AF173380.1 |
- |
4, 5 |
800 |
|
AF493235.1 |
- |
4, 5 |
865 |
|
AK030510.1 |
- |
4, 5 |
925 |
|
AK031693.1 |
- |
4, 5 |
895 |
|
AK035576.1 |
- |
2, 3 |
329 |
|
AY246699.1 |
- |
4, 5 |
623 |
|
AY626781.1 |
- |
4, 5 |
958 |
|
AY626782.1 |
- |
6, 7 |
946 |
|
AY626783.1 |
- |
7, 8 |
3164 |
|
BC041777.1 |
- |
7, 8 |
2007 |
4240274 |
BC042206.1 |
- |
7, 8 |
2007 |
4240274 |
BC043321.1 |
- |
6, 7 |
912 |
|
BC089009.1 |
- |
8, 9 |
3224 |
30543240 |
View Full Product Details