Gene Transcripts

Gene Symbol
Moap1
Entrez Gene ID
299261
Gene Name
modulator of apoptosis 1
Gene Aliases
Chromosome Location
Chr. 6: 126620597 - 126622532 on Build Rnor_6.0
UniGene ID
Rn.98234
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013101.1 NP_001013119.1 2 1762
GenBank mRNA BC091310.1 - 2 1762

Gene Symbol
LOC100233176
Entrez Gene ID
100233176
Gene Name
hypothetical protein LOC100233176
Gene Aliases
Chromosome Location
Chr. 6: 126620597 - 126636484 on Build Rnor_6.0
UniGene ID
Rn.98234
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142942.2 NP_001136414.1 2 1882

View Full Product Details