Gene Transcripts

Gene Symbol
Inpp5k
Entrez Gene ID
287533
Gene Name
inositol polyphosphate-5-phosphatase K
Gene Aliases
Skip
Chromosome Location
Chr. 10: 63776268 - 63795902 on Build Rnor_6.0
UniGene ID
Rn.7440
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013859.1 NP_001013881.1 10, 11 1291
GenBank mRNA BC083572.1 - 10, 11 1291
FQ228522.1 - 8, 9 2200

View Full Product Details