Gene Transcripts

Gene Symbol
Olr1225
Entrez Gene ID
405974
Gene Name
olfactory receptor 1225
Gene Aliases
Chromosome Location
Chr. 8: 41336340 - 41337323 on Build Rnor_6.0
UniGene ID
Rn.143543
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001083.1 NP_001001083.1 1 402

Gene Symbol
LOC100911999
Entrez Gene ID
100911999
Gene Name
olfactory receptor 143-like
Gene Aliases
Chromosome Location
Chr. 8: 41987837 - 41988832 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766103.1 XP_008764325.1 414

View Full Product Details