Gene Transcripts

Gene Symbol
Poll
Entrez Gene ID
361767
Gene Name
polymerase (DNA directed), lambda
Gene Aliases
Chromosome Location
Chr. 1: 265290337 - 265298840 on Build Rnor_6.0
UniGene ID
Rn.11799
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014168.1 NP_001014190.1 3 191
XM_006231545.2 XP_006231607.1 2 97
GenBank mRNA BC085841.1 - 3 191

View Full Product Details