Gene Transcripts

Gene Symbol
Tmem165
Entrez Gene ID
364137
Gene Name
transmembrane protein 165
Gene Aliases
Tparl
Chromosome Location
Chr. 14: 34503037 - 34528262 on Build Rnor_6.0
UniGene ID
Rn.2779
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024802.1 NP_001019973.1 5 1055
GenBank mRNA BC097478.1 - 5 1055

View Full Product Details