Gene Transcripts

Gene Symbol
Spatc1l
Entrez Gene ID
499418
Gene Name
spermatogenesis and centriole associated 1-like
Gene Aliases
Chromosome Location
Chr. 20: 12825789 - 12835007 on Build Rnor_6.0
UniGene ID
Rn.208235
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024296.1 NP_001019467.1 2 572
GenBank mRNA BC079291.1 - 2 572

View Full Product Details