Gene Transcripts

Gene Symbol
AIM1L
Entrez Gene ID
55057
Gene Name
absent in melanoma 1-like
Gene Aliases
CRYBG2
Chromosome Location
Chr. 1: 26321859 - 26354130 on Build GRCh38
UniGene ID
Hs.128738
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039775.3 NP_001034864.2 2 371
XM_005245918.2 XP_005245975.1 2 357
XM_011541672.1 XP_011539974.1 2 357
XM_011541673.2 XP_011539975.1 2 596
GenBank mRNA AK000902.1 - 2 371
DA439029.1 - 2 357

View Full Product Details