Gene Transcripts

Gene Symbol
Olr162
Entrez Gene ID
405295
Gene Name
olfactory receptor 162
Gene Aliases
Chromosome Location
Chr. 1: 169683311 - 169684270 on Build Rnor_6.0
UniGene ID
Rn.141841
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000950.1 NP_001000950.1 1 933

View Full Product Details