Gene Transcripts

Gene Symbol
MOB1A
Entrez Gene ID
55233
Gene Name
MOB kinase activator 1A
Gene Aliases
C2orf6, MATS1, MOB1, MOBK1B, MOBKL1B, Mob4B
Chromosome Location
Chr. 2: 74152598 - 74178967 on Build GRCh38
UniGene ID
Hs.602092
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317110.1 NP_001304039.1 6 1991
NM_001317111.1 NP_001304040.1 6 1887
NM_001317112.1 NP_001304041.1 5 1830
NM_018221.4 NP_060691.2 6 1994
XM_011532948.2 XP_011531250.1 5 1690
GenBank mRNA AK001650.1 - 6 1884
AK023321.1 - 6 1854

View Full Product Details