Gene Transcripts

Gene Symbol
ZFR2
Entrez Gene ID
23217
Gene Name
zinc finger RNA binding protein 2
Gene Aliases
KIAA1086
Chromosome Location
Chr. 19: 3804024 - 3869029 on Build GRCh38
UniGene ID
Hs.65750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145640.1 NP_001139112.1 2 789
NR_027149.1 3 947
GenBank mRNA AL365406.1 - 3 933
BC034005.1 - 2 793

View Full Product Details