Gene Transcripts

Gene Symbol
LRRC14B
Entrez Gene ID
389257
Gene Name
leucine rich repeat containing 14B
Gene Aliases
Chromosome Location
Chr. 5: 191511 - 195353 on Build GRCh38
UniGene ID
Hs.683662
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080478.1 NP_001073947.1 2 1377

View Full Product Details