Gene Transcripts
- Gene Symbol
- NAV2-AS4
- Entrez Gene ID
- 399876
- Gene Name
- NAV2 antisense RNA 4
- Gene Aliases
- Chromosome Location
- Chr. 11: 19510893 - 19524078 on Build GRCh38
- UniGene ID
- Hs.132600
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033989.1 |
|
4 |
601 |
|
GenBank mRNA |
AK023501.1 |
- |
4 |
601 |
|
View Full Product Details