Gene Transcripts

Gene Symbol
C5orf58
Entrez Gene ID
133874
Gene Name
chromosome 5 open reading frame 58
Gene Aliases
Chromosome Location
Chr. 5: 170231730 - 170252569 on Build GRCh38
UniGene ID
Hs.519749
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011534432.2 XP_011532734.1 1 371
XM_017009029.1 XP_016864518.1 1 1087
XM_017009030.1 XP_016864519.1 1 1086

View Full Product Details