Gene Transcripts

Gene Symbol
ABRACL
Entrez Gene ID
58527
Gene Name
ABRA C-terminal like
Gene Aliases
C6orf115, Costars, HSPC280, PRO2013
Chromosome Location
Chr. 6: 139028682 - 139043302 on Build GRCh38
UniGene ID
Hs.600861
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021243.2 NP_067066.1 1 75
GenBank mRNA AF161398.1 - 1 56
BG563686.1 - 1 76

View Full Product Details