Gene Transcripts
- Gene Symbol
- ANKRD20A5P
- Entrez Gene ID
- 440482
- Gene Name
- ankyrin repeat domain 20 family member A5, pseudogene
- Gene Aliases
- ANKRD20A5
- Chromosome Location
- Chr. 18: 14179097 - 14227050 on Build GRCh38
- UniGene ID
- Hs.632352
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC022023.1 |
- |
3 |
786 |
|
BC127726.1 |
- |
3 |
566 |
40127384 |
- Gene Symbol
- FLJ43315
- Entrez Gene ID
- 644316
- Gene Name
- asparagine synthetase pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 9: 64016950 - 64061612 on Build GRCh38
- UniGene ID
- Hs.632789
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC057848.1 |
- |
5 |
936 |
|
- Gene Symbol
- LOC102723552
- Entrez Gene ID
- 102723552
- Gene Name
- ankyrin repeat domain-containing protein 20A2
- Gene Aliases
- Chromosome Location
- Chr. 20: 30652373 - 30723916 on Build GRCh38
- UniGene ID
- Hs.741115
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC127725.1 |
- |
2 |
451 |
|
View Full Product Details