Gene Transcripts

Gene Symbol
Olfr1512
Entrez Gene ID
258424
Gene Name
olfactory receptor 1512
Gene Aliases
MOR223-5
Chromosome Location
Chr. 14: 52372110 - 52373051 on Build GRCm38
UniGene ID
Mm.377493
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146432.2 NP_666643.1 1 738
GenBank mRNA BC133656.1 - 1 738

View Full Product Details