Gene Transcripts

Gene Symbol
Olfr20
Entrez Gene ID
258925
Gene Name
olfactory receptor 20
Gene Aliases
MOR135-11, MTPCR06, MTPCR55, Olfr21
Chromosome Location
Chr. 11: 73350859 - 73354699 on Build GRCm38
UniGene ID
Mm.246526
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146923.2 NP_667134.2 2 135
GenBank mRNA BC138172.1 - 1 38
BC138173.1 - 1 38 8861192

View Full Product Details