Gene Transcripts

Gene Symbol
Olfr205
Entrez Gene ID
257881
Gene Name
olfactory receptor 205
Gene Aliases
GA_x54KRFPKG5P-55543875-55542958, GA_x5J8B7U0478-6-340, MOR182-11P
Chromosome Location
Chr. 16: 59328590 - 59329507 on Build GRCm38
UniGene ID
Mm.377282
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011736.1 NP_001011736.1 1 15
GenBank mRNA BC150839.1 - 1 40

View Full Product Details