Gene Transcripts

Gene Symbol
Olfr734
Entrez Gene ID
258658
Gene Name
olfactory receptor 734
Gene Aliases
MOR242-1
Chromosome Location
Chr. 14: 50304110 - 50320862 on Build GRCm38
UniGene ID
Mm.377621
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146664.1 NP_666875.1 1 557
GenBank mRNA BC141029.1 - 1 592

View Full Product Details