Gene Transcripts

Gene Symbol
Olfr206
Entrez Gene ID
258993
Gene Name
olfactory receptor 206
Gene Aliases
MOR182-4
Chromosome Location
Chr. 16: 59344779 - 59345699 on Build GRCm38
UniGene ID
Mm.377781
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146991.1 NP_667202.1 1 708
GenBank mRNA BC138396.1 - 1 735
BC138397.1 - 1 735 8861417

View Full Product Details