Gene Transcripts

Gene Symbol
NDUFB7
Entrez Gene ID
4713
Gene Name
NADH:ubiquinone oxidoreductase subunit B7
Gene Aliases
B18, CI-B18
Chromosome Location
Chr. 19: 14566078 - 14572077 on Build GRCh38
UniGene ID
Hs.532853
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004146.5 NP_004137.2 3 375
XM_011528039.2 XP_011526341.1 3 967
GenBank mRNA AF112200.1 - 3 361
AF217091.1 - 3 320
BC002595.2 - 3 360 3140806
BU565399.1 - 376
CB124371.1 - 3 375
CR450354.1 - 3 298
M33374.1 - 3 330

View Full Product Details