Gene Transcripts

Gene Symbol
ZNF439
Entrez Gene ID
90594
Gene Name
zinc finger protein 439
Gene Aliases
Chromosome Location
Chr. 19: 11866029 - 11869491 on Build GRCh38
UniGene ID
Hs.528731
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152262.2 NP_689475.1 3 1778
GenBank mRNA AK027542.1 - 1 1405
AL833935.1 - 3 1779
BC032857.1 - 3 1650 5268314

View Full Product Details