Gene Transcripts
- Gene Symbol
- AAGAB
- Entrez Gene ID
- 79719
- Gene Name
- alpha- and gamma-adaptin binding protein
- Gene Aliases
- KPPP1, PPKP1, PPKP1A, p34
- Chromosome Location
- Chr. 15: 67200675 - 67255198 on Build GRCh38
- UniGene ID
- Hs.254642
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271885.1 |
NP_001258814.1
|
4, 5 |
441 |
|
NM_001271886.1 |
NP_001258815.1
|
4, 5 |
702 |
|
NM_024666.4 |
NP_078942.3
|
4, 5 |
557 |
|
XM_011522020.1 |
XP_011520322.1
|
4, 5 |
807 |
|
GenBank mRNA |
AK021568.1 |
- |
4, 5 |
461 |
|
AK294402.1 |
- |
4, 5 |
438 |
|
AL136715.1 |
- |
4, 5 |
467 |
|
BC001975.1 |
- |
1, 2 |
78 |
|
BC047026.1 |
- |
4, 5 |
480 |
5534878 |
BC058886.1 |
- |
4, 5 |
479 |
6503238 |
CR533540.1 |
- |
4, 5 |
452 |
|
DC315715.1 |
- |
4, 5 |
441 |
|
View Full Product Details